Gene Transcripts
- Gene Symbol
- OOSP1
- Entrez Gene ID
- 255649
- Gene Name
- oocyte secreted protein 1, pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 11: 59942879 - 59995845 on Build GRCh38
- UniGene ID
- Hs.722773
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_130719.1 |
|
4, 5 |
552 |
|
GenBank mRNA |
AB231701.1 |
- |
1, 2 |
168 |
|
AB231702.1 |
- |
4, 5 |
549 |
|
View Full Product Details