Gene Transcripts
- Gene Symbol
- JMJD8
- Entrez Gene ID
- 339123
- Gene Name
- jumonji domain containing 8
- Gene Aliases
- C16orf20, PP14397
- Chromosome Location
- Chr. 16: 681666 - 684446 on Build GRCh38
- UniGene ID
- Hs.533771
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001005920.2 |
NP_001005920.2
|
2, 3 |
289 |
|
NM_001323918.1 |
NP_001310847.1
|
2, 3 |
296 |
|
NM_001323919.1 |
NP_001310848.1
|
2, 3 |
296 |
|
NR_136650.1 |
|
2, 3 |
296 |
|
NR_136651.1 |
|
2, 3 |
296 |
|
GenBank mRNA |
AF370420.1 |
- |
2, 3 |
173 |
|
AL524336.3 |
- |
2, 3 |
180 |
|
AL524922.3 |
- |
2, 3 |
183 |
|
BC073785.1 |
- |
1, 2 |
261 |
|
BC137100.1 |
- |
2, 3 |
255 |
9021097 |
BC137101.1 |
- |
2, 3 |
255 |
9021098 |
BG481039.1 |
- |
2, 3 |
192 |
|
BI463867.1 |
- |
2, 3 |
294 |
|
BX335015.2 |
- |
2, 3 |
176 |
|
CD674105.1 |
- |
2, 3 |
196 |
|
View Full Product Details