Gene Transcripts
- Gene Symbol
- SIMC1
- Entrez Gene ID
- 375484
- Gene Name
- SUMO interacting motifs containing 1
- Gene Aliases
- C5orf25, OOMA1, PLEIAD
- Chromosome Location
- Chr. 5: 176238359 - 176345991 on Build GRCh38
- UniGene ID
- Hs.719847
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001308195.1 |
NP_001295124.1
|
9, 10 |
2574 |
|
NM_001308196.1 |
NP_001295125.1
|
11, 12 |
2782 |
|
NM_001308200.1 |
NP_001295129.1
|
6, 7 |
1030 |
|
NM_198567.5 |
NP_940969.3
|
8, 9 |
1272 |
|
NR_131772.1 |
|
8, 9 |
1272 |
|
XM_011534553.2 |
XP_011532855.1
|
10, 11 |
6579 |
|
XM_011534554.2 |
XP_011532856.1
|
10, 11 |
2451 |
|
XM_011534556.2 |
XP_011532858.1
|
9, 10 |
6448 |
|
XM_017009454.1 |
XP_016864943.1
|
9, 10 |
2382 |
|
XM_017009455.1 |
XP_016864944.1
|
10, 11 |
2692 |
|
XM_017009456.1 |
XP_016864945.1
|
10, 11 |
2588 |
|
GenBank mRNA |
BC032390.1 |
- |
6, 7 |
998 |
|
BC037298.1 |
- |
11, 12 |
2783 |
4823991 |
BC066980.1 |
- |
8, 9 |
1265 |
4821958 |
BC073880.1 |
- |
6, 7 |
1030 |
|
KF151170.1 |
- |
9, 10 |
2442 |
|
View Full Product Details