Gene Transcripts

Gene Symbol
OR51Q1
Entrez Gene ID
390061
Gene Name
olfactory receptor family 51 subfamily Q member 1 (gene/pseudogene)
Gene Aliases
Chromosome Location
Chr. 11: 5422111 - 5423206 on Build GRCh38
UniGene ID
Hs.528592
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004757.2 NP_001004757.1 1 705
GenBank mRNA BC136999.1 - 1 705

View Full Product Details