Gene Transcripts
- Gene Symbol
- Barx2
- Entrez Gene ID
- 12023
- Gene Name
- BarH-like homeobox 2
- Gene Aliases
- 2310006E12Rik, Barx2b
- Chromosome Location
- Chr. 9: 31846044 - 31913285 on Build GRCm38
- UniGene ID
- Mm.261044
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013800.2 |
NP_038828.2
|
4 |
838 |
|
GenBank mRNA |
L77900.1 |
- |
4 |
636 |
|
View Full Product Details