Gene Transcripts

Gene Symbol
Cyp2c37
Entrez Gene ID
13096
Gene Name
cytochrome P450, family 2. subfamily c, polypeptide 37
Gene Aliases
Chromosome Location
Chr. 19: 39992424 - 40012243 on Build GRCm38
UniGene ID
Mm.38963
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010001.2 NP_034131.2 4, 5 663
GenBank mRNA AF047542.1 - 647
AK005017.1 - 4, 5 664
BC057912.1 - 648 5052873

View Full Product Details