Gene Transcripts

Gene Symbol
Diap1
Entrez Gene ID
13367
Gene Name
diaphanous homolog 1 (Drosophila)
Gene Aliases
D18Wsu154e, Dia1, Diaph1, Drf1, p140mDia
Chromosome Location
Chr. 18: 37843601 - 37935617 on Build GRCm38
UniGene ID
Mm.195916
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK140734.1 - 8, 9 881
AK220466.1 - 8, 9 890
BC021396.1 - 7, 8 920
BC070412.1 - 9, 10 1055 5705112
U96963.1 - 7, 8 893

View Full Product Details