Gene Transcripts

Gene Symbol
Cdca3
Entrez Gene ID
14793
Gene Name
cell division cycle associated 3
Gene Aliases
2410005A12Rik, C8, Grcc8, Tome-1
Chromosome Location
Chr. 6: 124829595 - 124833701 on Build GRCm38
UniGene ID
Mm.285723
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013538.5 NP_038566.1 4 554
GenBank mRNA AK008606.1 - 4 507
AK010409.1 - 4 495
AK011313.1 - 4 527
AK028159.1 - 4 496
AK145537.1 - 4 496
AK169792.1 - 4 502
BC002006.1 - 4 508 3592108
BC027172.1 - 3 1132

View Full Product Details