Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318186.1 | NP_001305115.1 | 15 | 1415 | |
NM_001318187.1 | NP_001305116.1 | 15 | 1415 | ||
NM_001318188.1 | NP_001305117.1 | 16 | 1487 | ||
NM_002140.4 | NP_002131.2 | 16 | 1648 | ||
NM_031262.3 | NP_112552.1 | 16 | 1648 | ||
NM_031263.3 | NP_112553.1 | 16 | 1487 | ||
XM_005251960.2 | XP_005252017.1 | 16 | 1466 | ||
XM_005251963.3 | XP_005252020.1 | 15 | 1428 | ||
XM_005251965.2 | XP_005252022.1 | 15 | 1386 | ||
XM_011518616.1 | XP_011516918.1 | 15 | 1428 | ||
XM_017014668.1 | XP_016870157.1 | 16 | 1466 | ||
XM_017014669.1 | XP_016870158.1 | 15 | 1386 | ||
GenBank mRNA | AB209562.1 | - | 15 | 1344 | |
AB451263.1 | - | 14 | 1261 | ||
AB451390.1 | - | 14 | 1261 | ||
AK096385.1 | - | 15 | 1273 | ||
AK123117.1 | - | 10 | 2213 | ||
AK291336.1 | - | 16 | 1470 | ||
AK294067.1 | - | 15 | 1445 | ||
AK300744.1 | - | 15 | 1358 | ||
AY911506.1 | - | 15 | 1324 | ||
BC000355.2 | - | 15 | 1383 | 2964383 | |
BC014980.1 | - | 16 | 1454 | 4906241 | |
BC025321.1 | - | 16 | 1406 | ||
CR456771.1 | - | 14 | 1261 | ||
S74678.1 | - | 15 | 1368 | ||
X72727.1 | - | 16 | 1470 |