Gene Transcripts

Gene Symbol
Inhbb
Entrez Gene ID
16324
Gene Name
inhibin beta-B
Gene Aliases
Chromosome Location
Chr. 1: 119415465 - 119422248 on Build GRCm38
UniGene ID
Mm.3092
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008381.3 NP_032407.1 2 1775
GenBank mRNA AK132352.1 - 2 1777
BC048845.1 - 2 481
X83376.1 - 1 146

View Full Product Details