Gene Transcripts

Gene Symbol
Slc12a3
Entrez Gene ID
20497
Gene Name
solute carrier family 12, member 3
Gene Aliases
AI035291, NCC, NCCT, TSC
Chromosome Location
Chr. 8: 94329112 - 94366222 on Build GRCm38
UniGene ID
Mm.25804
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001205311.1 NP_001192240.1 16 2033
NM_019415.2 NP_062288.2 16 2030
GenBank mRNA AK052691.1 - 16 2034
AK052755.1 - 16 2032
AK085496.1 - 16 2039
BC038612.1 - 16 2055 4237274
U61085.1 - 16 2012

View Full Product Details