Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009320.4 | NP_033346.2 | 11 | 1574 | |
GenBank mRNA | AK143510.1 | - | 11 | 1538 | |
AK185760.1 | - | 1 | 84 | ||
BC015245.1 | - | 11 | 1511 | 4242193 | |
L03292.2 | - | 11 | 1546 |