Gene Transcripts
- Gene Symbol
- Ube2k
- Entrez Gene ID
- 53323
- Gene Name
- ubiquitin-conjugating enzyme E2K
- Gene Aliases
- AW492011, D5Ertd601e, E2-25k, Hip2, Hypg, Lig
- Chromosome Location
- Chr. 5: 65537238 - 65598989 on Build GRCm38
- UniGene ID
- Mm.319512
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016786.3 |
NP_058066.2
|
5, 6 |
614 |
|
GenBank mRNA |
AB011081.1 |
- |
5, 6 |
552 |
|
AK006316.1 |
- |
2, 3 |
139 |
|
AK018366.1 |
- |
5, 6 |
587 |
|
AK031055.1 |
- |
3, 4 |
314 |
|
AK035371.1 |
- |
5, 6 |
511 |
|
AK036068.1 |
- |
5, 6 |
555 |
|
AK048194.1 |
- |
5, 6 |
605 |
|
AK139564.1 |
- |
5, 6 |
631 |
|
AK160728.1 |
- |
5, 6 |
574 |
|
AK162142.1 |
- |
5, 6 |
555 |
|
AK166647.1 |
- |
5, 6 |
523 |
|
BC002013.1 |
- |
5, 6 |
462 |
3496393 |
BC085311.1 |
- |
5, 6 |
540 |
30148009 |
View Full Product Details