Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001081695.2 | NP_001075164.1 | 5 | 509 | |
NM_001284197.1 | NP_001271126.1 | 3 | 350 | ||
NM_019448.4 | NP_062321.1 | 4 | 363 | ||
GenBank mRNA | AF220524.1 | - | 3 | 294 | |
AJ404467.1 | - | 3 | 335 | ||
AK010434.1 | - | 4 | 364 | ||
AV473569.1 | - | 3 | 350 | ||
BC083147.1 | - | 3 | 348 | 30447906 | |
EF051624.1 | - | 5 | 509 |