Gene Transcripts

Gene Symbol
Sult3a1
Entrez Gene ID
57430
Gene Name
sulfotransferase family 3A, member 1
Gene Aliases
ST3A1, Sult-x2, Sultx2
Chromosome Location
Chr. 10: 33851020 - 33879475 on Build GRCm38
UniGene ID
Mm.6590
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020565.2 NP_065590.2 6 800
GenBank mRNA AF026075.1 - 6 801
BC125593.1 - 6 800 40130108
BC125595.1 - 6 800 40130110

View Full Product Details