Gene Transcripts
- Gene Symbol
- Mrps21
- Entrez Gene ID
- 66292
- Gene Name
- mitochondrial ribosomal protein S21
- Gene Aliases
- 1810031B19Rik
- Chromosome Location
- Chr. 3: 95862646 - 95871604 on Build GRCm38
- UniGene ID
- Mm.24108
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_078479.3 |
NP_510964.1
|
1, 2 |
119 |
|
GenBank mRNA |
AB049956.1 |
- |
2, 3 |
144 |
|
AK002588.1 |
- |
2, 3 |
147 |
|
AK003462.1 |
- |
2, 3 |
140 |
|
AK003602.1 |
- |
2, 3 |
139 |
|
AK007662.1 |
- |
2, 3 |
148 |
|
AK007838.1 |
- |
1, 2 |
130 |
|
AK008265.1 |
- |
2, 3 |
162 |
|
AK008293.1 |
- |
2, 3 |
141 |
|
AK009049.1 |
- |
2, 3 |
140 |
|
AK009703.1 |
- |
2, 3 |
148 |
|
BC027546.1 |
- |
1, 2 |
123 |
1515797 |
BC125658.1 |
- |
1, 2 |
120 |
40130173 |
BC125660.1 |
- |
1, 2 |
120 |
40130175 |
View Full Product Details