Gene Transcripts

Gene Symbol
Nop9
Entrez Gene ID
67842
Gene Name
NOP9 nucleolar protein
Gene Aliases
2610027L16Rik
Chromosome Location
Chr. 14: 55745693 - 55755634 on Build GRCm38
UniGene ID
Mm.180338
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026403.3 NP_080679.3 9, 10 1831
GenBank mRNA AK011570.1 - 9, 10 1821
AK015585.1 - 9, 10 1581
AK032873.1 - 9, 10 1815
AK037831.1 - 9, 10 1819
AK050912.1 - 9, 10 1822
AK083861.1 - 9, 10 1700
AK139689.1 - 9, 10 1784
AK146020.1 - 9, 10 1786
AK146687.1 - 9, 10 1803
AK152732.1 - 9, 10 1805
AK154387.1 - 9, 10 1823
AK159798.1 - 9, 10 1805
BC018523.1 - 9, 10 1811 3499986

View Full Product Details