Gene Transcripts

Gene Symbol
Sncaip
Entrez Gene ID
67847
Gene Name
synuclein, alpha interacting protein (synphilin)
Gene Aliases
2810407O15Rik, 4933427B05Rik, BB104594, SYPH1
Chromosome Location
Chr. 18: 52767811 - 52915935 on Build GRCm38
UniGene ID
Mm.292168
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199151.1 NP_001186080.1 10 2338
NM_001199153.1 NP_001186082.1 10 2243
NM_001199154.1 NP_001186083.1 10 2513
NM_026408.4 NP_080684.3 10 2338
GenBank mRNA AF348447.1 - 10 2266
AK013040.1 - 8 2072
AK017012.1 - 7 1273
AK081942.1 - 10 2517
AK082079.1 - 10 2342
AK132346.1 - 10 2441
AK160111.1 - 10 2342
BC098489.1 - 10 2242 30096466
BC130225.1 - 10 2228
BC152544.1 - 10 2226

View Full Product Details