Gene Transcripts
- Gene Symbol
- Psmd11
- Entrez Gene ID
- 69077
- Gene Name
- proteasome (prosome, macropain) 26S subunit, non-ATPase, 11
- Gene Aliases
- 1700089D09Rik, 1810019E17Rik, 2610024G20Rik, 2810055C24Rik, C78232, P44.5, S9
- Chromosome Location
- Chr. 11: 80428615 - 80473248 on Build GRCm38
- UniGene ID
- Mm.260539
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_178616.3 |
NP_848731.2
|
11, 12 |
1135 |
|
GenBank mRNA |
AK007547.1 |
- |
11, 12 |
1104 |
|
AK012951.1 |
- |
10, 11 |
1599 |
|
AK029365.1 |
- |
7, 8 |
706 |
|
AK051738.1 |
- |
11, 12 |
1125 |
|
AK076002.1 |
- |
11, 12 |
1123 |
|
AK089942.1 |
- |
11, 12 |
1100 |
|
AK136049.1 |
- |
11, 12 |
1098 |
|
AK140773.1 |
- |
|
1120 |
|
AK167559.1 |
- |
11, 12 |
1106 |
|
AK168289.1 |
- |
11, 12 |
1127 |
|
BC030432.1 |
- |
8, 9 |
789 |
|
BC037590.1 |
- |
11, 12 |
1084 |
|
BC055457.1 |
- |
11, 12 |
1067 |
|
BC090664.1 |
- |
11, 12 |
1087 |
30677672 |
BC090980.1 |
- |
11, 12 |
1102 |
30283128 |
BC100595.1 |
- |
11, 12 |
1097 |
6767671 |
BC119136.1 |
- |
11, 12 |
1106 |
8733885 |
BC119138.1 |
- |
11, 12 |
1106 |
8733887 |
View Full Product Details