Gene Transcripts

Gene Symbol
Wdr34
Entrez Gene ID
71820
Gene Name
WD repeat domain 34
Gene Aliases
3200002I06Rik
Chromosome Location
Chr. 2: 30031551 - 30048879 on Build GRCm38
UniGene ID
Mm.88053
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008498.2 NP_001008498.2 4 584
GenBank mRNA AK014304.1 - 4 583
BC085113.1 - 4 540 30730999

View Full Product Details