Gene Transcripts

Gene Symbol
Nupl1
Entrez Gene ID
71844
Gene Name
nucleoporin like 1
Gene Aliases
1700017F11Rik, mKIAA0410
Chromosome Location
Chr. 14: 60205610 - 60251502 on Build GRCm38
UniGene ID
Mm.258051
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_170591.1 NP_733479.1 11 1280
GenBank mRNA AK006062.1 - 11 1383
AK029688.1 - 1334
AK078279.1 - 11 1328
AK086412.1 - 11 1348
AK129134.1 - 11 1278
AK133721.1 - 11 1291
AK139895.1 - 11 1411
AK146477.1 - 11 1280
BC026743.1 - 11 1311 3964978

View Full Product Details