Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001168535.1 | NP_001162007.1 | 5 | 883 | |
NM_001168536.1 | NP_001162008.1 | 5 | 883 | ||
NM_001168537.1 | NP_001162009.1 | 5 | 883 | ||
NM_001168538.1 | NP_001162010.1 | 5 | 883 | ||
NM_027986.3 | NP_082262.1 | 5 | 883 | ||
GenBank mRNA | AK009280.1 | - | 5 | 875 | |
AK032600.1 | - | 5 | 862 | ||
AK032813.1 | - | 5 | 869 | ||
AK039148.1 | - | 5 | 871 | ||
AK042494.1 | - | 5 | 872 | ||
AK045962.1 | - | 5 | 869 | ||
BC006901.1 | - | 5 | 845 | 3600168 |