Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028228.3 | NP_082504.1 | 5 | 491 | |
GenBank mRNA | AF321817.1 | - | 5 | 439 | |
AF421879.1 | - | 5 | 374 | ||
AJ344106.1 | - | 4 | 263 | ||
AK012057.1 | - | 5 | 469 | ||
AK075840.1 | - | 3 | 261 | ||
BC065781.1 | - | 5 | 390 | ||
BC115636.1 | - | 5 | 403 | 40102952 | |
BC115637.1 | - | 5 | 401 |