Gene Transcripts

Gene Symbol
Il23a
Entrez Gene ID
83430
Gene Name
interleukin 23, alpha subunit p19
Gene Aliases
IL-23, p19
Chromosome Location
Chr. 10: 128296140 - 128298084 on Build GRCm38
UniGene ID
Mm.125482
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031252.2 NP_112542.1 4 763
GenBank mRNA AF301619.1 - 4 763
AK043894.1 - 1650
BC019953.1 - 4 747 3708687

View Full Product Details