Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023668.2 | NP_076157.2 | 8 | 1009 | |
GenBank mRNA | AF290472.1 | - | 8 | 941 | |
AF323918.1 | - | 8 | 1029 | ||
AK011168.1 | - | 8 | 919 | ||
BC021434.1 | - | 905 | 5029832 | ||
BC046796.1 | - | 8 | 940 | 5718032 |