Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001130187.1 | NP_001123659.1 | 4 | 743 | |
NM_031878.2 | NP_114084.2 | 3 | 377 | ||
GenBank mRNA | AK159259.1 | - | 4 | 580 | |
AK166123.1 | - | 4 | 536 | ||
BC005732.1 | - | 3 | 367 | 2647494 | |
CO425905.1 | - | 3 | 544 |