Gene Transcripts
- Gene Symbol
- Hmox2
- Entrez Gene ID
- 15369
- Gene Name
- heme oxygenase 2
- Gene Aliases
- HO-2, HO2
- Chromosome Location
- Chr. 16: 4726361 - 4766249 on Build GRCm38
- UniGene ID
- Mm.272866
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001136066.2 |
NP_001129538.1
|
5, 6 |
965 |
|
NM_010443.2 |
NP_034573.2
|
5, 6 |
981 |
|
GenBank mRNA |
AF029874.1 |
- |
5, 6 |
978 |
|
AF054669.1 |
- |
5, 6 |
982 |
|
AF054670.1 |
- |
5, 6 |
916 |
|
AK032175.1 |
- |
5, 6 |
923 |
|
AK134536.1 |
- |
5, 6 |
940 |
|
AK141426.1 |
- |
5, 6 |
959 |
|
AK152936.1 |
- |
5, 6 |
904 |
|
BC002011.1 |
- |
5, 6 |
937 |
3484259 |
View Full Product Details