Gene Transcripts

Gene Symbol
Olr1292
Entrez Gene ID
405312
Gene Name
olfactory receptor 1292
Gene Aliases
Chromosome Location
Chr. 15: 36286785 - 36287696 on Build Rnor_6.0
UniGene ID
Rn.143498
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000960.1 NP_001000960.1 1 287

Gene Symbol
LOC100910719
Entrez Gene ID
100910719
Gene Name
olfactory receptor 144-like
Gene Aliases
Chromosome Location
Chr. 15: 35586351 - 35587262 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008770724.1 XP_008768946.1 1 287

View Full Product Details