Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 58193302-58193551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNASE1L3
Entrez Gene ID
1776
Gene Name
deoxyribonuclease I like 3
Gene Aliases
DHP2, DNAS1L3, LSD, SLEB16
UniGene
Hs.476453
Gene Chromosome Location
Chr.3: 58192626-58211003 on Build GRCh38
Transcripts
NM_001256560.1 NM_004944.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3702465 c.760G>A p.V254I Chr.3: 58193384-58193384 on Build GRCh38
5918057 c.737C>T p.S246F Chr.3: 58193407-58193407 on Build GRCh38
1204233 c.724G>T p.E242* Chr.3: 58193420-58193420 on Build GRCh38
2782634 c.793G>A p.E265K Chr.3: 58193351-58193351 on Build GRCh38
5941792 c.724G>A p.E242K Chr.3: 58193420-58193420 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563541630 Chr.3: 58193333-58193333 on Build GRCh38
rs367807030 Chr.3: 58193353-58193353 on Build GRCh38
rs778387758 Chr.3: 58193414-58193414 on Build GRCh38
rs923622995 Chr.3: 58193529-58193529 on Build GRCh38
rs367994383 Chr.3: 58193477-58193477 on Build GRCh38
rs768456171 Chr.3: 58193425-58193425 on Build GRCh38
rs76440799 Chr.3: 58193415-58193415 on Build GRCh38
rs377465977 Chr.3: 58193329-58193329 on Build GRCh38
rs759275938 Chr.3: 58193466-58193466 on Build GRCh38
rs35044617 Chr.3: 58193528-58193528 on Build GRCh38
rs752793169 Chr.3: 58193386-58193386 on Build GRCh38
rs148314913 Chr.3: 58193380-58193380 on Build GRCh38
rs1035809592 Chr.3: 58193504-58193504 on Build GRCh38
rs373633551 Chr.3: 58193326-58193326 on Build GRCh38
rs776438281 Chr.3: 58193450-58193450 on Build GRCh38
rs760584857 Chr.3: 58193357-58193357 on Build GRCh38
rs763090079 Chr.3: 58193487-58193487 on Build GRCh38
rs146444966 Chr.3: 58193362-58193362 on Build GRCh38
rs778660483 Chr.3: 58193426-58193426 on Build GRCh38
rs138248740 Chr.3: 58193367-58193367 on Build GRCh38
rs774002626 Chr.3: 58193336-58193336 on Build GRCh38
rs369909357 Chr.3: 58193508-58193508 on Build GRCh38
rs186823038 Chr.3: 58193442-58193442 on Build GRCh38
rs193030547 Chr.3: 58193478-58193478 on Build GRCh38
rs906582065 Chr.3: 58193327-58193327 on Build GRCh38
rs764209862 Chr.3: 58193447-58193453 on Build GRCh38
rs746168066 Chr.3: 58193416-58193416 on Build GRCh38
rs759754450 Chr.3: 58193368-58193368 on Build GRCh38
rs761368740 Chr.3: 58193337-58193337 on Build GRCh38
rs545672132 Chr.3: 58193404-58193404 on Build GRCh38
rs772989463 Chr.3: 58193355-58193355 on Build GRCh38
rs747698984 Chr.3: 58193435-58193435 on Build GRCh38
rs760864502 Chr.3: 58193331-58193331 on Build GRCh38
rs752724246 Chr.3: 58193453-58193453 on Build GRCh38
rs760542731 Chr.3: 58193448-58193448 on Build GRCh38
rs770832574 Chr.3: 58193449-58193449 on Build GRCh38
rs76455752 Chr.3: 58193531-58193531 on Build GRCh38

View Full Product Details