Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 58205400-58205633 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNASE1L3
Entrez Gene ID
1776
Gene Name
deoxyribonuclease I like 3
Gene Aliases
DHP2, DNAS1L3, LSD, SLEB16
UniGene
Hs.476453
Gene Chromosome Location
Chr.3: 58192626-58211003 on Build GRCh38
Transcripts
NM_001256560.1 NM_004944.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5865166 c.301C>T p.Q101* Chr.3: 58205490-58205490 on Build GRCh38
7171241 c.298G>C p.E100Q Chr.3: 58205493-58205493 on Build GRCh38
6647457 c.251C>T p.T84M Chr.3: 58205540-58205540 on Build GRCh38
5585037 c.239G>A p.R80K Chr.3: 58205552-58205552 on Build GRCh38
4002798 c.252G>A p.T84T Chr.3: 58205539-58205539 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759756597 Chr.3: 58205580-58205580 on Build GRCh38
rs747361058 Chr.3: 58205432-58205432 on Build GRCh38
rs763068123 Chr.3: 58205600-58205600 on Build GRCh38
rs17058970 Chr.3: 58205515-58205515 on Build GRCh38
rs765457252 Chr.3: 58205594-58205594 on Build GRCh38
rs770698995 Chr.3: 58205552-58205552 on Build GRCh38
rs748939790 Chr.3: 58205533-58205533 on Build GRCh38
rs188393900 Chr.3: 58205569-58205569 on Build GRCh38
rs561644406 Chr.3: 58205516-58205516 on Build GRCh38
rs74350392 Chr.3: 58205547-58205547 on Build GRCh38
rs12491947 Chr.3: 58205503-58205503 on Build GRCh38
rs766912371 Chr.3: 58205464-58205467 on Build GRCh38
rs778293446 Chr.3: 58205540-58205540 on Build GRCh38
rs769594461 Chr.3: 58205433-58205433 on Build GRCh38
rs772902080 Chr.3: 58205473-58205473 on Build GRCh38
rs779923120 Chr.3: 58205531-58205531 on Build GRCh38
rs2070117 Chr.3: 58205539-58205539 on Build GRCh38
rs751206379 Chr.3: 58205500-58205501 on Build GRCh38
rs747642606 Chr.3: 58205543-58205543 on Build GRCh38
rs370250390 Chr.3: 58205447-58205447 on Build GRCh38
rs774409070 Chr.3: 58205457-58205457 on Build GRCh38
rs141477807 Chr.3: 58205517-58205517 on Build GRCh38
rs200875129 Chr.3: 58205485-58205485 on Build GRCh38
rs569181582 Chr.3: 58205602-58205602 on Build GRCh38
rs754628701 Chr.3: 58205591-58205593 on Build GRCh38
rs771981459 Chr.3: 58205470-58205470 on Build GRCh38
rs761672713 Chr.3: 58205461-58205461 on Build GRCh38
rs905697034 Chr.3: 58205460-58205460 on Build GRCh38
rs757664587 Chr.3: 58205530-58205530 on Build GRCh38
rs1036778398 Chr.3: 58205500-58205500 on Build GRCh38
rs746868593 Chr.3: 58205549-58205549 on Build GRCh38
rs370578082 Chr.3: 58205430-58205430 on Build GRCh38
rs143383107 Chr.3: 58205529-58205529 on Build GRCh38
rs375098703 Chr.3: 58205443-58205443 on Build GRCh38
rs376951611 Chr.3: 58205454-58205454 on Build GRCh38
rs999691545 Chr.3: 58205581-58205581 on Build GRCh38
rs144928501 Chr.3: 58205506-58205506 on Build GRCh38
rs775456424 Chr.3: 58205597-58205597 on Build GRCh38
rs779831079 Chr.3: 58205436-58205436 on Build GRCh38

View Full Product Details