Gene Transcripts

Gene Symbol
Adck3
Entrez Gene ID
67426
Gene Name
aarF domain containing kinase 3
Gene Aliases
4632432J16Rik, AI462003, Cabc1, mKIAA0451
Chromosome Location
Chr. 1: 180165238 - 180196020 on Build GRCm38
UniGene ID
Mm.38330
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163290.1 NP_001156762.1 11 1327
NM_023341.3 NP_075830.2 11 1425
GenBank mRNA AJ278735.1 - 11 1305
AK004177.1 - 4 310
AK004791.1 - 11 1367
AK014605.2 - 11 1367
AK191635.1 - 2 50
AK192575.1 - 2 50
AK194890.1 - 2 50
AK196005.1 - 2 50
AK199890.1 - 50
AK200253.1 - 2 50
AK202430.1 - 2 50
AK207786.1 - 2 50
AK217974.1 - 3 165
BC030937.1 - 11 1373 4922892
U31629.1 - 9 966

View Full Product Details