Gene Transcripts

Gene Symbol
Chchd5
Entrez Gene ID
66170
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 5
Gene Aliases
1110027L01Rik, AW045710
Chromosome Location
Chr. 2: 129129700 - 129134112 on Build GRCm38
UniGene ID
Mm.40621
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025395.3 NP_079671.1 2 261
GenBank mRNA AK003341.1 - 2 163
AK003936.1 - 2 144
AK013018.1 - 2 140
BC094414.1 - 2 152 1515083
BC096668.1 - 2 159 1530928
BY335381.1 - 2 264

View Full Product Details