Gene Transcripts

Gene Symbol
Mfap2
Entrez Gene ID
17150
Gene Name
microfibrillar-associated protein 2
Gene Aliases
AI893631, Magp, Magp1
Chromosome Location
Chr. 4: 141010424 - 141015984 on Build GRCm38
UniGene ID
Mm.7386
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161799.1 NP_001155271.1 3, 4 226
NM_008546.3 NP_032572.2 3, 4 252
GenBank mRNA AK160213.1 - 3, 4 228
AK164920.1 - 3, 4 209
BC035490.1 - 3, 4 186 5249342
CK389335.1 - 3, 4 252
DV654229.1 - 3, 4 226
L23769.1 - 3, 4 208

View Full Product Details