Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019990.4 | NP_064374.1 | 2 | 250 | |
GenBank mRNA | AB031550.1 | - | 3 | 427 | |
BC046335.1 | - | 2 | 339 | ||
BC058773.1 | - | 2 | 382 | ||
BC080808.1 | - | 2 | 445 | ||
BC090390.1 | - | 1 | 144 | ||
BC094007.1 | - | 2 | 263 | ||
BC116984.1 | - | 1 | 158 | 40126303 | |
BC120641.1 | - | 1 | 158 | 40129564 | |
BC144877.1 | - | 1 | 158 | ||
BG078855.2 | - | 1 | 36 | ||
BY079536.1 | - | 2 | 256 | ||
CJ070178.1 | - | 1 | 183 |