Gene Transcripts

Gene Symbol
Dcpp1
Entrez Gene ID
13184
Gene Name
demilune cell and parotid protein 1
Gene Aliases
BC005655, C86550, Dcpp, Dcpp-1, p20
Chromosome Location
Chr. 17: 23880876 - 23882853 on Build GRCm38
UniGene ID
Mm.371556
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC024627.1 - 210
BC045522.1 - 4 220 4190591
BC090399.1 - 210 4168483
DQ237936.1 - 228
S76879.1 - 269
U03711.1 - 233

View Full Product Details