Gene Transcripts

Gene Symbol
Plin1
Entrez Gene ID
103968
Gene Name
perilipin 1
Gene Aliases
6030432J05Rik, Peri, Plin
Chromosome Location
Chr. 7: 79721164 - 79732904 on Build GRCm38
UniGene ID
Mm.254917
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113471.1 NP_001106942.1 3 162
NM_175640.2 NP_783571.2 3 218
GenBank mRNA AK031445.1 - 2 143
AY161165.1 - 2 113
BC025137.1 - 3 278
BC096685.1 - 3 163 1448913
BY723040.1 - 3 277

View Full Product Details