Gene Transcripts

Gene Symbol
Podxl2
Entrez Gene ID
319655
Gene Name
podocalyxin-like 2
Gene Aliases
D130074J02Rik, PODLX2
Chromosome Location
Chr. 6: 88842557 - 88875053 on Build GRCm38
UniGene ID
Mm.302613
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176973.4 NP_795947.3 2 263
GenBank mRNA AK038943.1 - 2 295
AK051761.1 - 2 465
BC033384.1 - 2 263 5362581

View Full Product Details