Gene Transcripts

Gene Symbol
STK40
Entrez Gene ID
83931
Gene Name
serine/threonine kinase 40
Gene Aliases
SHIK, SgK495
Chromosome Location
Chr. 1: 36339619 - 36385927 on Build GRCh38
UniGene ID
Hs.471768
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282546.1 NP_001269475.1 5 751
NM_001282547.1 NP_001269476.1 5 736
NM_032017.2 NP_114406.1 6 973
GenBank mRNA AK024504.1 - 4 535
AK075029.1 - 2 252
AK075048.1 - 5 733
AL834137.1 - 6 930
AY198395.1 - 5 706
BC007835.2 - 5 704 4109652
BC008344.1 - 5 712 3530483

View Full Product Details