Gene Transcripts
- Gene Symbol
- LONP2
- Entrez Gene ID
- 83752
- Gene Name
- lon peptidase 2, peroxisomal
- Gene Aliases
- LONP, LONPL
- Chromosome Location
- Chr. 16: 48244167 - 48353979 on Build GRCh38
- UniGene ID
- Hs.295923
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001300948.1 |
NP_001287877.1
|
3, 4 |
820 |
|
NM_031490.3 |
NP_113678.2
|
4, 5 |
952 |
|
XM_017023755.1 |
XP_016879244.1
|
4, 5 |
819 |
|
XM_017023756.1 |
XP_016879245.1
|
3, 4 |
685 |
|
GenBank mRNA |
AJ548761.1 |
- |
4, 5 |
819 |
|
AK027544.1 |
- |
4, 5 |
783 |
|
AK074775.1 |
- |
4, 5 |
817 |
|
AK127125.1 |
- |
4, 5 |
584 |
|
AK294779.1 |
- |
3, 4 |
632 |
|
AL834201.1 |
- |
3, 4 |
319 |
|
BC093910.1 |
- |
4, 5 |
786 |
7939755 |
BC093912.1 |
- |
4, 5 |
786 |
7939757 |
BC143246.1 |
- |
3, 4 |
654 |
9051747 |
View Full Product Details