Gene Transcripts

Gene Symbol
ADAM33
Entrez Gene ID
80332
Gene Name
ADAM metallopeptidase domain 33
Gene Aliases
C20orf153, DJ964F7.1
Chromosome Location
Chr. 20: 3667973 - 3682131 on Build GRCh38
UniGene ID
Hs.173716
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282447.2 NP_001269376.1 6 1171
NM_025220.4 NP_079496.1 6 1171
NM_153202.3 NP_694882.1 6 1171
XM_005260843.1 XP_005260900.1 6 628
XM_006723639.1 XP_006723702.1 6 628
XM_006723640.1 XP_006723703.1 6 628
XM_006723644.2 XP_006723707.1 6 628
XM_011529366.1 XP_011527668.1 6 527
XM_011529367.1 XP_011527669.1 6 488
XM_011529368.1 XP_011527670.1 6 628
XM_011529370.1 XP_011527672.1 6 628
XM_011529371.1 XP_011527673.1 6 628
XM_011529373.1 XP_011527675.1 6 525
XM_017028080.1 XP_016883569.1 6 628
XM_017028081.1 XP_016883570.1 6 549
XM_017028082.1 XP_016883571.1 6 628
XM_017028083.1 XP_016883572.1 6 628
GenBank mRNA AB055891.1 - 6 572
AF466287.1 - 6 490
AK123015.1 - 3 651
AK126487.1 - 4 1427
AK126498.1 - 4 333
AK226148.1 - 5 875
AK300429.1 - 6 539
AK304037.1 - 6 525
AY358314.1 - 6 598
BC125112.1 - 4 788
BC125113.1 - 6 532 40118831
DB225158.1 - 6 549

View Full Product Details