Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003374.2 | NP_003365.1 | 9 | 1296 | |
NR_036624.1 | 9 | 1374 | |||
NR_036625.1 | 9 | 1377 | |||
XM_005272075.3 | XP_005272132.1 | 9 | 1293 | ||
XM_017009821.1 | XP_016865310.1 | 9 | 1224 | ||
XM_017009822.1 | XP_016865311.1 | 9 | 1203 | ||
XM_017009823.1 | XP_016865312.1 | 9 | 1226 | ||
GenBank mRNA | AK095989.1 | - | 9 | 1296 | |
AK122953.1 | - | 12 | 2699 | ||
AK293639.1 | - | 7 | 753 | ||
BC008482.1 | - | 9 | 1081 | 4332199 | |
BC071168.1 | - | 9 | 1113 | 6023095 | |
BC090042.1 | - | 9 | 1106 | 30395350 | |
L06132.1 | - | 9 | 1150 |