Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012471.2 | NP_036603.1 | 8 | 3005 | |
XM_017029774.1 | XP_016885263.1 | 9 | 3295 | ||
GenBank mRNA | AB209258.1 | - | 8 | 2419 | |
AF054568.1 | - | 8 | 3004 | ||
BC137271.1 | - | 8 | 2845 | 9021268 | |
BC137274.1 | - | 8 | 2845 | 9021271 |