Gene Transcripts

Gene Symbol
MTMR4
Entrez Gene ID
9110
Gene Name
myotubularin related protein 4
Gene Aliases
FYVE-DSP2, ZFYVE11
Chromosome Location
Chr. 17: 58489532 - 58518109 on Build GRCh38
UniGene ID
Hs.514373
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004687.4 NP_004678.3 7 612
XM_005257784.2 XP_005257841.1 6 761
XM_005257785.4 XP_005257842.1 8 729
XM_005257786.4 XP_005257843.1 7 1516
XM_006722168.3 XP_006722231.1 7 556
XM_011525460.2 XP_011523762.1 8 1609
GenBank mRNA BC035609.1 - 7 573

View Full Product Details