Gene Transcripts
- Gene Symbol
- SCYL1
- Entrez Gene ID
- 57410
- Gene Name
- SCY1 like pseudokinase 1
- Gene Aliases
- GKLP, HT019, NKTL, NTKL, P105, SCAR21, TAPK, TEIF, TRAP
- Chromosome Location
- Chr. 11: 65525065 - 65538711 on Build GRCh38
- UniGene ID
- Hs.238839
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001048218.1 |
NP_001041683.1
|
6, 7 |
925 |
|
NM_020680.3 |
NP_065731.3
|
6, 7 |
925 |
|
XM_005274118.3 |
XP_005274175.1
|
6, 7 |
937 |
|
XM_005274120.3 |
XP_005274177.1
|
6, 7 |
937 |
|
XM_005274121.3 |
XP_005274178.1
|
6, 7 |
937 |
|
XM_017018029.1 |
XP_016873518.1
|
6, 7 |
937 |
|
XM_017018030.1 |
XP_016873519.1
|
6, 7 |
937 |
|
XM_017018031.1 |
XP_016873520.1
|
6, 7 |
937 |
|
XM_017018032.1 |
XP_016873521.1
|
6, 7 |
779 |
|
XM_017018033.1 |
XP_016873522.1
|
6, 7 |
929 |
|
GenBank mRNA |
AB047077.1 |
- |
6, 7 |
848 |
|
AB051427.1 |
- |
6, 7 |
848 |
|
AB051428.1 |
- |
6, 7 |
848 |
|
AF297709.1 |
- |
6, 7 |
887 |
|
AK314945.1 |
- |
6, 7 |
881 |
|
BC069233.1 |
- |
6, 7 |
889 |
6581110 |
View Full Product Details