Gene Transcripts

Gene Symbol
SPCS3
Entrez Gene ID
60559
Gene Name
signal peptidase complex subunit 3
Gene Aliases
PRO3567, SPC22/23, SPC3, YLR066W
Chromosome Location
Chr. 4: 176319939 - 176332245 on Build GRCh38
UniGene ID
Hs.42194
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021928.3 NP_068747.1 5 837
GenBank mRNA AK026302.1 - 5 811
AK092634.1 - 4 1819
BC047290.1 - 5 781 5224927

View Full Product Details