Gene Transcripts
- Gene Symbol
- KCNH6
- Entrez Gene ID
- 81033
- Gene Name
- potassium voltage-gated channel subfamily H member 6
- Gene Aliases
- ERG-2, ERG2, HERG2, Kv11.2, hERG-2
- Chromosome Location
- Chr. 17: 63523334 - 63548977 on Build GRCh38
- UniGene ID
- Hs.591177
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001278919.1 |
NP_001265848.1
|
1, 2 |
165 |
|
NM_001278920.1 |
NP_001265849.1
|
1, 2 |
165 |
|
NM_030779.3 |
NP_110406.1
|
1, 2 |
165 |
|
NM_173092.2 |
NP_775115.1
|
1, 2 |
165 |
|
XM_011525308.2 |
XP_011523610.1
|
1, 2 |
165 |
|
XM_011525309.2 |
XP_011523611.1
|
1, 2 |
165 |
|
XM_011525310.2 |
XP_011523612.1
|
1, 2 |
165 |
|
XM_011525311.1 |
XP_011523613.1
|
1, 2 |
165 |
|
XM_011525312.1 |
XP_011523614.1
|
1, 2 |
165 |
|
XM_011525313.1 |
XP_011523615.1
|
1, 2 |
165 |
|
XM_017025175.1 |
XP_016880664.1
|
1, 2 |
165 |
|
XM_017025176.1 |
XP_016880665.1
|
1, 2 |
165 |
|
XM_017025177.1 |
XP_016880666.1
|
1, 2 |
165 |
|
XM_017025178.1 |
XP_016880667.1
|
1, 2 |
165 |
|
XM_017025179.1 |
XP_016880668.1
|
1, 2 |
165 |
|
XM_017025180.1 |
XP_016880669.1
|
1, 2 |
134 |
|
GenBank mRNA |
AF311913.1 |
- |
1, 2 |
96 |
|
AK091877.1 |
- |
1, 2 |
165 |
|
AK296498.1 |
- |
1, 2 |
138 |
|
AK298363.1 |
- |
1, 2 |
140 |
|
BC006334.2 |
- |
1, 2 |
143 |
4303609 |
DB147394.1 |
- |
1, 2 |
138 |
|
DB506421.1 |
- |
1, 2 |
360 |
|
View Full Product Details