Gene Transcripts
- Gene Symbol
- KLK5
- Entrez Gene ID
- 25818
- Gene Name
- kallikrein related peptidase 5
- Gene Aliases
- KLK-L2, KLKL2, SCTE
- Chromosome Location
- Chr. 19: 50943299 - 50953088 on Build GRCh38
- UniGene ID
- Hs.50915
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001077491.1 |
NP_001070959.1
|
3, 4 |
301 |
|
NM_001077492.1 |
NP_001070960.1
|
2, 3 |
233 |
|
NM_012427.4 |
NP_036559.1
|
2, 3 |
436 |
|
XM_011526702.1 |
XP_011525004.1
|
2, 3 |
175 |
|
XM_011526703.2 |
XP_011525005.1
|
3, 4 |
255 |
|
GenBank mRNA |
AF168768.2 |
- |
2, 3 |
252 |
|
AF435980.1 |
- |
2, 3 |
407 |
|
AF435981.1 |
- |
2, 3 |
204 |
|
AK223421.1 |
- |
2, 3 |
384 |
|
AY279380.1 |
- |
2, 3 |
210 |
|
AY279381.1 |
- |
3, 4 |
278 |
|
AY359010.1 |
- |
2, 3 |
436 |
|
AY461805.1 |
- |
1, 2 |
94 |
|
BC008036.2 |
- |
2, 3 |
367 |
2989806 |
BT006867.1 |
- |
1, 2 |
83 |
|
View Full Product Details