Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001871.2 | NP_001862.2 | 4 | 368 | |
GenBank mRNA | AJ224866.1 | - | 4 | 350 | |
AK313940.1 | - | 4 | 356 | ||
BC015338.1 | - | 4 | 349 | 4422620 | |
BI770145.1 | - | 4 | 368 | ||
BT009910.1 | - | 4 | 345 | ||
M81057.1 | - | 4 | 368 |