Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001007026.1 | NP_001007027.1 | 5 | 2501 | |
NM_001940.3 | NP_001931.2 | 5 | 2494 | ||
GenBank mRNA | BC051795.2 | - | 5 | 2515 | |
BQ880124.1 | - | 1 | 157 | ||
D31840.1 | - | 5 | 2487 | ||
D38529.1 | - | 4 | 2417 | ||
U23851.1 | - | 4 | 2319 |