Gene Transcripts
- Gene Symbol
- ARID4A
- Entrez Gene ID
- 5926
- Gene Name
- AT-rich interaction domain 4A
- Gene Aliases
- RBBP-1, RBBP1, RBP-1, RBP1
- Chromosome Location
- Chr. 14: 58298385 - 58373733 on Build GRCh38
- UniGene ID
- Hs.161000
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002892.3 |
NP_002883.3
|
13, 14 |
1321 |
|
NM_023000.2 |
NP_075376.2
|
13, 14 |
1321 |
|
NM_023001.2 |
NP_075377.2
|
13, 14 |
1321 |
|
XM_005267964.2 |
XP_005268021.1
|
13, 14 |
1440 |
|
XM_005267965.2 |
XP_005268022.1
|
13, 14 |
1440 |
|
XM_017021560.1 |
XP_016877049.1
|
13, 14 |
1440 |
|
XM_017021561.1 |
XP_016877050.1
|
13, 14 |
1217 |
|
XM_017021562.1 |
XP_016877051.1
|
13, 14 |
1440 |
|
XM_017021563.1 |
XP_016877052.1
|
13, 14 |
1440 |
|
XM_017021564.1 |
XP_016877053.1
|
11, 12 |
1048 |
|
XM_017021565.1 |
XP_016877054.1
|
3, 4 |
1483 |
|
GenBank mRNA |
AF086075.1 |
- |
1, 2 |
68 |
|
AK307520.1 |
- |
13, 14 |
1270 |
|
AK307972.1 |
- |
|
299 |
|
BC026230.1 |
- |
13, 14 |
1269 |
|
S57153.1 |
- |
1, 2 |
56 |
|
S57160.1 |
- |
1, 2 |
56 |
|
S57162.1 |
- |
1, 2 |
56 |
|
S66427.1 |
- |
13, 14 |
1181 |
|
View Full Product Details